Product Details

SNP ID
rs28798168
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:14113697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGATGAAAACTATAAACCCACT[A/G]TATTAGTCTGTTCTCACACTGCTAT
Phenotype
MIM: 609252
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LIPI PubMed Links

Gene Details

Gene
LIPI
Gene Name
lipase I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302998.1 Intron NP_001289927.1
NM_001302999.1 Intron NP_001289928.1
NM_001303000.1 Intron NP_001289929.1
NM_001303001.1 Intron NP_001289930.1
NM_198996.3 Intron NP_945347.2
XM_006723965.3 Intron XP_006724028.1

View Full Product Details