Product Details

SNP ID
rs34924859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:39520615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTTCTCTGCGCGTGTGCGTTTT[C/T]CCTCCTCCCCGCCCTCAGGGTCCAC
Phenotype
MIM: 604064 MIM: 615497 MIM: 610225
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATF4 PubMed Links

Gene Details

Gene
ATF4
Gene Name
activating transcription factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001675.4 531 Intron NP_001666.2
NM_182810.2 531 Intron NP_877962.1
XM_017028807.1 531 UTR 5 XP_016884296.1
Gene
MIEF1
Gene Name
mitochondrial elongation factor 1
There are no transcripts associated with this gene.

Gene
RPS19BP1
Gene Name
ribosomal protein S19 binding protein 1
There are no transcripts associated with this gene.

View Full Product Details