Product Details

SNP ID
rs28549096
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50557562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATACTGTTCAGCACAGTACCTACC[A/G]TAATGAGCACTTAATAAATCATATC
Phenotype
MIM: 615775
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHDC7B PubMed Links

Gene Details

Gene
KLHDC7B
Gene Name
kelch domain containing 7B
There are no transcripts associated with this gene.

Gene
SYCE3
Gene Name
synaptonemal complex central element protein 3
There are no transcripts associated with this gene.

View Full Product Details