Product Details

SNP ID
rs28602639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:56987634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTAATCACTATTGGGCAAATCCA[C/T]CAAAAAGTAGCTGAATGAATGAATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPIN3 PubMed Links

Gene Details

Gene
SPIN3
Gene Name
spindlin family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010862.2 Intron NP_001010862.2
XM_006724577.3 Intron XP_006724640.1
XM_006724578.3 Intron XP_006724641.1
XM_017029315.1 Intron XP_016884804.1

View Full Product Details