Product Details

SNP ID
rs28382148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:71367104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGAGTGAAGCTGTCGGCCGCTGT[C/G]GCCCCTCCTGTGCCGCAGGCGCATG
Phenotype
MIM: 313650
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TAF1 PubMed Links

Gene Details

Gene
TAF1
Gene Name
TATA-box binding protein associated factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286074.1 Intron NP_001273003.1
NM_004606.4 Intron NP_004597.2
NM_138923.3 Intron NP_620278.1
XM_005262295.1 Intron XP_005262352.1
XM_005262296.1 Intron XP_005262353.1
XM_005262297.4 Intron XP_005262354.1
XM_005262300.2 Intron XP_005262357.1
XM_006724682.3 Intron XP_006724745.1

View Full Product Details