Product Details

SNP ID
rs28643457
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:76325743 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGTTTAAATTACACAGAATCAGT[G/T]AGAGCTTACACATAAGAGCAGCATG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC158 PubMed Links

Gene Details

Gene
CCDC158
Gene Name
coiled-coil domain containing 158
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042784.1 Intron NP_001036249.1
XM_005262974.4 Intron XP_005263031.1
XM_006714204.3 Intron XP_006714267.1
XM_011531908.2 Intron XP_011530210.1
XM_011531909.2 Intron XP_011530211.1
XM_011531910.2 Intron XP_011530212.1
XM_011531911.1 Intron XP_011530213.1
XM_011531912.2 Intron XP_011530214.1
XM_011531913.1 Intron XP_011530215.1
XM_011531914.2 Intron XP_011530216.1
XM_011531915.2 Intron XP_011530217.1
XM_011531916.2 Intron XP_011530218.1
XM_011531917.1 Intron XP_011530219.1
XM_017008160.1 Intron XP_016863649.1
XM_017008161.1 Intron XP_016863650.1

View Full Product Details