Product Details

SNP ID
rs28713721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:67914142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATGCTTTTTGTCTTTTCTATAAC[A/G]TTATTTTCTTATTCTGAAATTGGAT
Phenotype
MIM: 611704
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMPRSS11A PubMed Links

Gene Details

Gene
TMPRSS11A
Gene Name
transmembrane protease, serine 11A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114387.1 Intron NP_001107859.1
NM_182606.3 Intron NP_872412.3

View Full Product Details