Product Details

SNP ID
rs28610062
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:88618907 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCTTTCTAGTTCTTAAAAGTAA[C/T]GTGTGATAAGGCCTCAAATAGATTT
Phenotype
MIM: 613279
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EML5 PubMed Links

Gene Details

Gene
EML5
Gene Name
echinoderm microtubule associated protein like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_183387.2 Intron NP_899243.1
XM_006720070.3 Intron XP_006720133.1
XM_011536528.2 Intron XP_011534830.1
XM_011536530.2 Intron XP_011534832.1
XM_011536531.2 Intron XP_011534833.1
XM_011536532.2 Intron XP_011534834.1
XM_011536533.2 Intron XP_011534835.1
XM_011536534.2 Intron XP_011534836.1
XM_011536536.2 Intron XP_011534838.1
XM_011536538.2 Intron XP_011534840.1
XM_017021062.1 Intron XP_016876551.1
XM_017021063.1 Intron XP_016876552.1
XM_017021064.1 Intron XP_016876553.1
XM_017021065.1 Intron XP_016876554.1
XM_017021066.1 Intron XP_016876555.1
XM_017021067.1 Intron XP_016876556.1
XM_017021068.1 Intron XP_016876557.1
XM_017021069.1 Intron XP_016876558.1
XM_017021070.1 Intron XP_016876559.1
Gene
ZC3H14
Gene Name
zinc finger CCCH-type containing 14
There are no transcripts associated with this gene.

View Full Product Details