Product Details
- SNP ID
-
rs11540290
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52470517 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCTCACTGTACCTGCTTCTTGAC[A/G]TGGTCGATCTCAGATCTCAGCCTCT
- Phenotype
-
MIM: 612315
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT6C
PubMed Links
Gene Details
- Gene
- KRT6C
- Gene Name
- keratin 6C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173086.4 |
1239 |
Silent Mutation |
CAC,CAT |
H397H |
NP_775109.2 |
View Full Product Details