Product Details

SNP ID
rs28688234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62544762 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGGCAGAAGGCGAAGGAGGAGCA[A/G]GCTTGTTACATTGCCAGAGCCGGAA
Phenotype
MIM: 609326
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C20orf166-AS1 PubMed Links

Gene Details

Gene
C20orf166-AS1
Gene Name
C20orf166 antisense RNA 1
There are no transcripts associated with this gene.

Gene
MIR1-1
Gene Name
microRNA 1-1
There are no transcripts associated with this gene.

Gene
MIR1-1HG
Gene Name
MIR1-1 host gene
There are no transcripts associated with this gene.

View Full Product Details