Product Details
- SNP ID
-
rs28683753
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:1364857 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCGAGCATCTCCCACACGAGCAGG[A/G]CCAGGGTGGTCACGGGTGCTGGGGA
- Phenotype
-
MIM: 607838
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GNPTG
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74823949,rs80296423] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GNPTG
- Gene Name
- N-acetylglucosamine-1-phosphate transferase gamma subunit
There are no transcripts associated with this gene.
- Gene
- UNKL
- Gene Name
- unkempt family like zinc finger
View Full Product Details