Product Details

SNP ID
rs28447317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2030540 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGCGGCCTGGCAGGCCTTGGGG[A/G]CCAGGAGGAACAGCCTGAAGCTGTC
Phenotype
MIM: 607997 MIM: 602656 MIM: 606553
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NPW PubMed Links

Gene Details

Gene
NPW
Gene Name
neuropeptide W
There are no transcripts associated with this gene.

Gene
NTHL1
Gene Name
nth-like DNA glycosylase 1
There are no transcripts associated with this gene.

Gene
SLC9A3R2
Gene Name
SLC9A3 regulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130012.2 Intron NP_001123484.1
NM_001252073.1 Intron NP_001239002.1
NM_001252075.1 Intron NP_001239004.1
NM_001252076.1 Intron NP_001239005.1
NM_004785.5 Intron NP_004776.3
XM_005255702.4 Intron XP_005255759.1
XM_006720981.1 Intron XP_006721044.1
XM_017023894.1 Intron XP_016879383.1
XM_017023895.1 Intron XP_016879384.1

View Full Product Details