Product Details
- SNP ID
-
rs28420645
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:70519450 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCTAGGTGGACTTCTCTTTGACTG[C/T]TCAAGATGGTTCAAGACCATTAAGA
- Phenotype
-
MIM: 610912
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AMTN
PubMed Links
Gene Details
- Gene
- AMTN
- Gene Name
- amelotin
View Full Product Details