Product Details

SNP ID
rs61730141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65794053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCAGCCTGGGCTTCTGCCCACC[A/G]CAGCCCTACCGGGAGCCGGAACCCT
Phenotype
MIM: 602313
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OVOL1 PubMed Links

Gene Details

Gene
OVOL1
Gene Name
ovo like transcriptional repressor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004561.3 463 Silent Mutation CCA,CCG P41P NP_004552.2
XM_005274018.3 463 UTR 5 XP_005274075.1
XM_011545067.2 463 UTR 5 XP_011543369.1
XM_011545068.2 463 UTR 5 XP_011543370.1
XM_017017837.1 463 UTR 5 XP_016873326.1
XM_017017838.1 463 UTR 5 XP_016873327.1
Gene
OVOL1-AS1
Gene Name
OVOL1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details