Product Details

SNP ID
rs141017859
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:631279 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCCCGCCCTACGTCCACTCCTGC[C/T]GCTCCTGCTCCTCCTCCGGCTGACC
Phenotype
MIM: 608021
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LOC100287175 PubMed Links
Additional Information
For this assay, SNP(s) [rs4984905,rs8062289] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LOC100287175
Gene Name
uncharacterized LOC100287175
There are no transcripts associated with this gene.

Gene
METTL26
Gene Name
methyltransferase like 26
There are no transcripts associated with this gene.

Gene
RAB40C
Gene Name
RAB40C, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
WFIKKN1
Gene Name
WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053284.2 268 Missense Mutation CCG,CTG P9L NP_444514.1

View Full Product Details