Product Details

SNP ID
rs41258845
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:45888992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGAGTCTGACGCCTCCGCTCCC[C/G]ATGCTGCAGGTACCACAGTCCCGCC
Phenotype
MIM: 146730
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IGFBP1 PubMed Links

Gene Details

Gene
IGFBP1
Gene Name
insulin like growth factor binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000596.2 633 Missense Mutation CAT,GAT H114D NP_000587.1

View Full Product Details