Product Details
- SNP ID
-
hCV86412372
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:153950898 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAGATTTCGCTGAAGGGCCCCCGG[C/G]CACAGGCATTGATTCCGGCAACACG
- Phenotype
-
MIM: 300019
MIM: 312420
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
HCFC1
PubMed Links
Gene Details
- Gene
- HCFC1
- Gene Name
- host cell factor C1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005334.2 |
6146 |
Missense Mutation |
GCC,GGC |
A1873G |
NP_005325.2 |
- Gene
- RENBP
- Gene Name
- renin binding protein
There are no transcripts associated with this gene.
View Full Product Details