Product Details

SNP ID
rs41261245
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:119240266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGTGTATAAATCACAGAGAAAT[A/T]GACTGTCTAATAAAGAATTGATCTT
Phenotype
MIM: 300435
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC107985692 PubMed Links

Gene Details

Gene
LOC107985692
Gene Name
A-kinase anchor protein 17B-like
There are no transcripts associated with this gene.

Gene
PGRMC1
Gene Name
progesterone receptor membrane component 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282621.1 Intron NP_001269550.1
NM_006667.4 Intron NP_006658.1

View Full Product Details