Product Details

SNP ID
rs56212137
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:163143855 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATGCTTATTAGTTTATTAATTTA[C/T]TTGTTAATGGCCCAGTTTTTATTGA
Phenotype
MIM: 603276
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RGS5 PubMed Links

Gene Details

Gene
RGS5
Gene Name
regulator of G-protein signaling 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195303.2 4371 UTR 3 NP_001182232.1
NM_001254748.1 4371 UTR 3 NP_001241677.1
NM_001254749.1 4371 UTR 3 NP_001241678.1
NM_003617.3 4371 UTR 3 NP_003608.1

View Full Product Details