Product Details
- SNP ID
-
rs61309556
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:191199161 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGTGGAAAATCTGTAGAAATGAT[G/T]TAAGAGAAAGCAAAGGCAGGGATTG
- Phenotype
-
MIM: 610280
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
OSTN
PubMed Links
Gene Details
- Gene
- OSTN
- Gene Name
- osteocrin
View Full Product Details