Product Details

SNP ID
rs62017905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:72481234 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTATGGCAGTAGTCCTATGAATCA[A/G]ACTCTGGGGATGGGGTCTGGCTGTC
Phenotype
MIM: 605624
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARIH1 PubMed Links

Gene Details

Gene
ARIH1
Gene Name
ariadne RBR E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005744.3 Intron NP_005735.2
XM_011521438.2 Intron XP_011519740.1
XM_011521439.2 Intron XP_011519741.1
Gene
LOC105370888
Gene Name
uncharacterized LOC105370888
There are no transcripts associated with this gene.

View Full Product Details