Product Details

SNP ID
rs58360980
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1723184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGAATCATGTTGCTTTGGTGCC[A/G]GCTGTCTGGTTTCTATTCCATATCC
Phenotype
MIM: 612077 MIM: 614218
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MIR22 PubMed Links

Gene Details

Gene
MIR22
Gene Name
microRNA 22
There are no transcripts associated with this gene.

Gene
MIR22HG
Gene Name
MIR22 host gene
There are no transcripts associated with this gene.

Gene
WDR81
Gene Name
WD repeat domain 81
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163673.1 Intron NP_001157145.1
NM_001163809.1 Intron NP_001157281.1
NM_001163811.1 Intron NP_001157283.1
NM_152348.3 Intron NP_689561.2
XM_011523651.2 Intron XP_011521953.1
XM_017024184.1 Intron XP_016879673.1

View Full Product Details