Product Details

SNP ID
rs58392883
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:32597187 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTATCACAAGGTCATTTGAAAA[C/T]GGACTCAGGACAAACCCATATACGT
Phenotype
MIM: 604583
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD27 PubMed Links

Gene Details

Gene
ANKRD27
Gene Name
ankyrin repeat domain 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032139.2 4267 UTR 3 NP_115515.2
XM_006723412.2 4267 UTR 3 XP_006723475.1
XM_017027349.1 4267 UTR 3 XP_016882838.1
Gene
PDCD5
Gene Name
programmed cell death 5
There are no transcripts associated with this gene.

View Full Product Details