Product Details

SNP ID
rs58755664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2715909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACACCTCACACATATCCAGTCTTG[C/T]TGACAACCACACACAGGTATCTACA
Phenotype
MIM: 607862
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DIRAS1 PubMed Links

Gene Details

Gene
DIRAS1
Gene Name
DIRAS family GTPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145173.3 2221 UTR 3 NP_660156.1
XM_011527713.2 2221 UTR 3 XP_011526015.1

View Full Product Details