Product Details

SNP ID
rs62123687
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17797785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTACACATGGTTTCACTAAAGCAG[G/T]GTCCGGCCATTGTGAACTTAAAGGA
Phenotype
MIM: 605863 MIM: 613437
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
B3GNT3 PubMed Links

Gene Details

Gene
B3GNT3
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014256.3 Intron NP_055071.2
XM_011527626.2 Intron XP_011525928.1
Gene
FCHO1
Gene Name
FCH domain only 1
There are no transcripts associated with this gene.

View Full Product Details