Product Details

SNP ID
rs59578783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:56536194 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAGCAGAGTCCTCCTGGCCTTC[C/G]AGGCTTCCCATTTCGCACTCCACTA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM209A PubMed Links

Gene Details

Gene
FAM209A
Gene Name
family with sequence similarity 209 member A
There are no transcripts associated with this gene.

Gene
FAM209B
Gene Name
family with sequence similarity 209 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013646.3 Intron NP_001013668.2
XM_017027849.1 Intron XP_016883338.1

View Full Product Details