Product Details

SNP ID
rs60411156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37706066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGAGATGGAGGCAAATGGACAC[A/G]TTTCCGAACTAATTGAGCAGGCACG
Phenotype
MIM: 609761
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIOBP PubMed Links

Gene Details

Gene
TRIOBP
Gene Name
TRIO and F-actin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039141.2 Intron NP_001034230.1
NM_007032.5 Intron NP_008963.3
NM_138632.2 Intron NP_619538.2

View Full Product Details