Product Details

SNP ID
rs56031201
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:39223809 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCGACTTTACAAACTGAAGGAA[C/G]CAGGTTTTGGAAGGCGGGAAGGGGG
Phenotype
MIM: 190040
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PDGFB PubMed Links

Gene Details

Gene
PDGFB
Gene Name
platelet derived growth factor subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002608.3 3351 UTR 3 NP_002599.1
NM_033016.3 3351 UTR 3 NP_148937.1

View Full Product Details