Product Details

SNP ID
rs59866166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:44569408 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGATTTTCCAGCTGGCAGAAAT[A/G]TCCTCTACCTTGAGGGAGGCGTGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00207 PubMed Links

Gene Details

Gene
LINC00207
Gene Name
long intergenic non-protein coding RNA 207
There are no transcripts associated with this gene.

View Full Product Details