Product Details

SNP ID
rs56828380
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:119748515 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAACACATGACAGGAAAGTTTATC[G/T]GGATCTTGAAGGCAACAGCTGATCT
Phenotype
MIM: 600234 MIM: 606879
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HMGCS2 PubMed Links

Gene Details

Gene
HMGCS2
Gene Name
3-hydroxy-3-methylglutaryl-CoA synthase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166107.1 1947 UTR 3 NP_001159579.1
NM_005518.3 1947 UTR 3 NP_005509.1
XM_011541313.1 1947 UTR 3 XP_011539615.1
Gene
PHGDH
Gene Name
phosphoglycerate dehydrogenase
There are no transcripts associated with this gene.

View Full Product Details