Product Details

SNP ID
rs59198366
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:69398896 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTACCCAAGGATGAGTGATTCCCA[A/G]TGGAGCCCTTAAGGGCACAGAATTA
Phenotype
MIM: 608100
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NFU1 PubMed Links

Gene Details

Gene
NFU1
Gene Name
NFU1 iron-sulfur cluster scaffold
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002755.2 Intron NP_001002755.1
NM_001002756.2 Intron NP_001002756.1
NM_015700.3 Intron NP_056515.2
XM_017003808.1 Intron XP_016859297.1

View Full Product Details