Product Details

SNP ID
rs61066015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:35047903 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAATGTTAGTAAGACATTTCCCC[C/T]AAACTGTGAGTATATCTCTGTGTAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM177A1 PubMed Links

Gene Details

Gene
FAM177A1
Gene Name
family with sequence similarity 177 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079519.1 Intron NP_001072987.1
NM_001289022.1 Intron NP_001275951.1
NM_173607.3 Intron NP_775878.2

View Full Product Details