Product Details

SNP ID
rs56363123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:129899932 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTAAGAGTTTTTTTGAAAGGAT[A/G]CTTCCAAGTCAGAAAACAAGAAGAT
Phenotype
MIM: 164013
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NFRKB PubMed Links

Gene Details

Gene
NFRKB
Gene Name
nuclear factor related to kappaB binding protein
There are no transcripts associated with this gene.

Gene
PRDM10
Gene Name
PR domain 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020228.2 7856 UTR 3 NP_064613.2
NM_199437.1 7856 UTR 3 NP_955469.1
NM_199438.1 7856 UTR 3 NP_955470.1
NM_199439.1 7856 UTR 3 NP_955471.1
XM_011542912.2 7856 UTR 3 XP_011541214.1
XM_017018023.1 7856 UTR 3 XP_016873512.1

View Full Product Details