Product Details

SNP ID
rs61759492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63974791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCGCCGGAGGGGAAGCTTGGGAT[A/C]ATGGTGAGGAACGGGCCTGGAAGAG
Phenotype
MIM: 123870
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
COX8A PubMed Links

Gene Details

Gene
COX8A
Gene Name
cytochrome c oxidase subunit 8A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004074.2 185 Silent Mutation ATA,ATC I37I NP_004065.1

View Full Product Details