Product Details
- SNP ID
-
rs62408132
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:5998115 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATCTCTCTCTCTTTTTTTTTTAAA[A/T]TCAAGGCTCTTTTATGTCAAAATCT
- Phenotype
-
MIM: 607409
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NRN1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3834316,rs564045003] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NRN1
- Gene Name
- neuritin 1
View Full Product Details