Product Details

SNP ID
rs59280971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:652196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCAAAATTCAAAAACTAAAGCAG[C/T]GTTTTTCCTGCAAGTGGGCGCACTG
Phenotype
MIM: 611380
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DIP2C PubMed Links

Gene Details

Gene
DIP2C
Gene Name
disco interacting protein 2 homolog C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014974.2 Intron NP_055789.1
XM_005252426.3 Intron XP_005252483.1
XM_005252427.3 Intron XP_005252484.1
XM_005252428.4 Intron XP_005252485.1
XM_005252430.2 Intron XP_005252487.2
XM_011519428.2 Intron XP_011517730.1
XM_011519429.2 Intron XP_011517731.1
XM_011519430.2 Intron XP_011517732.1
XM_011519431.2 Intron XP_011517733.1
XM_011519432.1 Intron XP_011517734.1
XM_011519433.2 Intron XP_011517735.1
XM_017015977.1 Intron XP_016871466.1
Gene
PRR26
Gene Name
proline rich 26
There are no transcripts associated with this gene.

View Full Product Details