Product Details

SNP ID
rs74144508
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:228510380 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCTCAGTGATTGTGAGGACAGAA[C/T]TGAATAGAGGACTCACCGGGCAGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BTNL10 PubMed Links

Gene Details

Gene
BTNL10
Gene Name
butyrophilin like 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287262.1 759 Silent Mutation CAA,CAG Q202Q NP_001274191.1
XM_011544015.2 759 Intron XP_011542317.1
XM_016999986.1 759 Intron XP_016855475.1
XM_016999987.1 759 Silent Mutation CAA,CAG Q202Q XP_016855476.1

View Full Product Details