Product Details

SNP ID
rs72763977
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:228510952 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCTTGTCTGTTTCGAAGCACAA[A/G]TTCTGAGTCACTCGGGAAACCTCCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BTNL10 PubMed Links

Gene Details

Gene
BTNL10
Gene Name
butyrophilin like 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287262.1 Intron NP_001274191.1
XM_011544015.2 Intron XP_011542317.1
XM_016999986.1 Intron XP_016855475.1
XM_016999987.1 Intron XP_016855476.1

View Full Product Details