Product Details

SNP ID
rs72725038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35407953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGTGCCTACTCATTACCCAGTGC[A/G]GCGTCAAAGCATGGTGAAGTAGTAG
Phenotype
MIM: 605836
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP8B5P PubMed Links

Gene Details

Gene
ATP8B5P
Gene Name
ATPase phospholipid transporting 8B5, pseudogene
There are no transcripts associated with this gene.

Gene
UNC13B
Gene Name
unc-13 homolog B
There are no transcripts associated with this gene.

View Full Product Details