Product Details

SNP ID
rs73976628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:9828066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTCTAAACCCCTCCAATCCCCAT[A/G]TAGGGCAGGCTGTGGTACCAGGTCT
Phenotype
MIM: 603659
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLP2R PubMed Links

Gene Details

Gene
GLP2R
Gene Name
glucagon like peptide 2 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004246.2 Intron NP_004237.1
XM_005256861.3 Intron XP_005256918.1
XM_011524077.2 Intron XP_011522379.1
XM_017025339.1 Intron XP_016880828.1
XM_017025340.1 Intron XP_016880829.1
XM_017025341.1 Intron XP_016880830.1
Gene
GSG1L2
Gene Name
GSG1 like 2
There are no transcripts associated with this gene.

View Full Product Details