Product Details

SNP ID
rs73554721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38730920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACAGTGGCTTGAGGCAGGGAGCT[C/T]GCAGGACAGAGCCTGAGCCACCCTG
Phenotype
MIM: 604638 MIM: 608839
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTN4 PubMed Links

Gene Details

Gene
ACTN4
Gene Name
actinin alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322033.1 Intron NP_001308962.1
NM_004924.5 Intron NP_004915.2
XM_005259281.4 Intron XP_005259338.1
XM_006723406.2 Intron XP_006723469.1
XM_017027331.1 Intron XP_016882820.1
Gene
CAPN12
Gene Name
calpain 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144691.4 Intron NP_653292.2
XM_017026355.1 Intron XP_016881844.1
XM_017026356.1 Intron XP_016881845.1
XM_017026357.1 Intron XP_016881846.1
XM_017026358.1 Intron XP_016881847.1
XM_017026359.1 Intron XP_016881848.1
XM_017026360.1 Intron XP_016881849.1
XM_017026361.1 Intron XP_016881850.1
XM_017026362.1 Intron XP_016881851.1
XM_017026363.1 Intron XP_016881852.1
XM_017026364.1 Intron XP_016881853.1
XM_017026365.1 Intron XP_016881854.1

View Full Product Details