Product Details

SNP ID
rs73878488
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:22486864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTGCTCTCCCTCAACTAACCTC[C/T]GCCTTCTTGGTCTCTGCTCACGCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF280B PubMed Links

Gene Details

Gene
ZNF280B
Gene Name
zinc finger protein 280B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080764.3 2944 UTR 3 NP_542942.2
XM_011529893.2 2944 UTR 3 XP_011528195.1
XM_011529895.2 2944 UTR 3 XP_011528197.1
XM_011529896.2 2944 UTR 3 XP_011528198.1
XM_011529897.2 2944 Intron XP_011528199.1

View Full Product Details