Product Details
- SNP ID
-
rs73437199
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:88493594 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTACTTTAGATTTCTTATCCCACT[G/T]TCCAATTTTAAAGTGTAATTAATGA
- Phenotype
-
MIM: 184745
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KITLG
PubMed Links
Gene Details
- Gene
- KITLG
- Gene Name
- KIT ligand
View Full Product Details