Product Details

SNP ID
rs74057656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:58251892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTTTATACTTACATATTAATAT[A/G]TTTTTAGTCAAATGATGTACTAGAT
Phenotype
MIM: 176843
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSMA3 PubMed Links

Gene Details

Gene
PSMA3
Gene Name
proteasome subunit alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002788.3 Intron NP_002779.1
NM_152132.2 Intron NP_687033.1

View Full Product Details