Product Details

SNP ID
rs73213995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196469175 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTGAGGCATTCAAGAATGTCCA[C/T]CCTAAAACTGAAAGTGAAATTGCTC
Phenotype
MIM: 612688
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF168 PubMed Links

Gene Details

Gene
RNF168
Gene Name
ring finger protein 168
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152617.3 4955 UTR 3 NP_689830.2

View Full Product Details