Product Details

SNP ID
rs80203913
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:180154993 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTCGCGGTTCCCGGCGCTAACG[C/T]GGCCCCGCGGTCGGCGCTCTATTCG
Phenotype
MIM: 603120
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
QSOX1 PubMed Links

Gene Details

Gene
QSOX1
Gene Name
quiescin sulfhydryl oxidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004128.2 161 Missense Mutation GCG,GTG A29V NP_001004128.1
NM_002826.4 161 Missense Mutation GCG,GTG A29V NP_002817.2

View Full Product Details