Product Details

SNP ID
rs76005585
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:154569471 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTCCCCTGCCCAGGGGTGTGGG[C/G]TACGGATACAGAGGAGCGGCTGGTG
Phenotype
MIM: 118507
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CHRNB2 PubMed Links

Gene Details

Gene
CHRNB2
Gene Name
cholinergic receptor nicotinic beta 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000748.2 320 Missense Mutation GCT,GGT A25G NP_000739.1
XM_017000180.1 320 UTR 5 XP_016855669.1

View Full Product Details