Product Details

SNP ID
rs78805380
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:25821809 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACTAGGGCCAGAGCTCTGACCT[C/T]ATCTGGCCACCTGCCTAATGCTACT
Phenotype
MIM: 606210
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC646471 PubMed Links

Gene Details

Gene
LOC646471
Gene Name
uncharacterized LOC646471
There are no transcripts associated with this gene.

Gene
MTFR1L
Gene Name
mitochondrial fission regulator 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099625.1 Intron NP_001093095.1
NM_001099626.1 Intron NP_001093096.1
NM_001099627.1 Intron NP_001093097.1
NM_019557.5 Intron NP_062457.3
Gene
SEPN1
Gene Name
selenoprotein N, 1
There are no transcripts associated with this gene.

View Full Product Details