Product Details

SNP ID
rs78203611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:149041249 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAACATCTGAAGGTAGGTCGTTC[A/G]AAGTGGCCACAGCTCTTTGGTGGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LYPD6B PubMed Links

Gene Details

Gene
LYPD6B
Gene Name
LY6/PLAUR domain containing 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317002.1 Intron NP_001303931.1
NM_001317003.1 Intron NP_001303932.1
NM_001317004.1 Intron NP_001303933.1
NM_001317005.1 Intron NP_001303934.1
NM_001317006.1 Intron NP_001303935.1
NM_177964.4 Intron NP_808879.2
XM_005246306.4 Intron XP_005246363.1
XM_006712279.2 Intron XP_006712342.1
XM_006712280.3 Intron XP_006712343.1
XM_006712281.3 Intron XP_006712344.1
XM_011510620.2 Intron XP_011508922.1
XM_011510623.2 Intron XP_011508925.1
XM_011510625.1 Intron XP_011508927.1
XM_017003364.1 Intron XP_016858853.1
XM_017003365.1 Intron XP_016858854.1
XM_017003366.1 Intron XP_016858855.1

View Full Product Details