Product Details

SNP ID
rs79736763
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:163609369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCATCTGGGGCTTTCAAATCAG[A/C]AGCTCTCATTTGATGCACTTTTCCT
Phenotype
MIM: 605295
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FIGN PubMed Links

Gene Details

Gene
FIGN
Gene Name
fidgetin, microtubule severing factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321825.1 2778 UTR 3 NP_001308754.1
NM_018086.3 2778 UTR 3 NP_060556.2
XM_011511389.2 2778 UTR 3 XP_011509691.1
XM_017004419.1 2778 UTR 3 XP_016859908.1

View Full Product Details